MAT428345657 Matrix Scientific

6-Methoxy-2-(4-nitrophenyl)-1,3-benzothiazole

  • MDL number MFCD06410799
  • Cas Number : 43036-14-2
  • Molecular Weight : 286.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient
Country of Origin US
Product Link http://www.matrixscientific.com/115028.html

Safety Information

MSDS http://www.matrixscientific.com/115028.html
Availability details for
Quantity
  • Availability
  • Add to Cart