MAT428345872 Matrix Scientific

1,4-Dihydrazinyl-phthalazine

  • MDL number MFCD01250766
  • Cas Number : 484-23-1
  • Molecular Weight : 190.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/115247.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/115247.html
Availability details for
Quantity
  • Availability
  • Add to Cart