MAT428345876 Matrix Scientific

(R)-2-(1-Naphthylmethyl)succinic acid-1-methyl ester

  • MDL number MFCD06798981
  • Cas Number : 119807-82-8
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/115251.html

Safety Information

MSDS http://www.matrixscientific.com/115251.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart