MAT428346498 Matrix Scientific

3-(Chloromethyl)-3-methyloxetane

  • MDL number MFCD01709552
  • Cas Number : 822-48-0
  • Molecular Weight : 120.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Density 1.1
Country of Origin USA
Boiling point 88
Product Link http://www.matrixscientific.com/115870.html

Safety Information

hcodes Flammable
MSDS http://www.matrixscientific.com/115870.html
Availability details for
Quantity
  • Availability
  • Add to Cart