MAT428346859 Matrix Scientific

Ethyl 5-(chlorosulfonyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD08691284
  • Cas Number : 115703-22-5
  • Molecular Weight : 238.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/116241.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/116241.html
Availability details for
Quantity
  • Availability
  • Add to Cart