MAT428499958 Matrix Scientific

2,5-Difluoro-DL-phenylglycine

  • MDL number MFCD02662504
  • Cas Number : 243448-17-1
  • Molecular Weight : 187.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/116479.html

Safety Information

MSDS http://www.matrixscientific.com/116479.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart