MAT428500078 Matrix Scientific

2-Fluoro-4-methylphenylacetic acid

  • MDL number MFCD03094329
  • Cas Number : 518070-28-5
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/116599.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/116599.html
Availability details for
Quantity
  • Availability
  • Add to Cart