MAT428500460 Matrix Scientific

3-Chloro-5-(trifluoromethoxy)phenylacetic acid

  • MDL number MFCD06660313
  • Cas Number : 433938-70-6
  • Molecular Weight : 254.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin US
Product Link https://www.matrixscientific.com/116981.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart