MAT428500586 Matrix Scientific

6-Chloro-2-fluoro-3-methoxyphenylacetic acid

  • MDL number MFCD09832273
  • Cas Number : 1017777-83-1
  • Molecular Weight : 218.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-137
Storage temp. ambient
Country of Origin US
Product Link https://www.matrixscientific.com/117107.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart