MAT428501104 Matrix Scientific

3-Fluoro-5-(trifluoromethoxy)phenylacetic acid

  • MDL number MFCD18783228
  • Cas Number : 1352999-94-0
  • Molecular Weight : 238.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/117625.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/117625.html
Availability details for
Quantity
  • Availability
  • Add to Cart