MAT428501197 Matrix Scientific

3-Methoxy-4-(trifluoromethyl)benzaldehyde

  • MDL number MFCD11110997
  • Cas Number : 945003-38-3
  • Molecular Weight : 204.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/117718.html

Safety Information

hcodes Irritant/Air Sensitive
MSDS http://www.matrixscientific.com/117718.html
Availability details for
Quantity
  • Availability
  • Add to Cart