MAT428501198 Matrix Scientific

3-Methoxy-4-(trifluoromethyl)benzoyl chloride

  • MDL number MFCD18398587
  • Cas Number : 1261571-92-9
  • Molecular Weight : 238.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/117719.html

Safety Information

hcodes Corrosive/Stored under Argon/Moisture Sensitive
MSDS http://www.matrixscientific.com/117719.html
Availability details for
Quantity
  • Availability
  • Add to Cart