MAT432291096 Matrix Scientific

3,5-Dichloro-1-[(4-methoxyphenyl)-methyl]-1,2-dihydropyrazin-2-one

  • MDL number MFCD24683345
  • Cas Number : 393860-82-7
  • Molecular Weight : 285.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-100
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/118755.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/118755.html
Availability details for
Quantity
  • Availability
  • Add to Cart