MAT432291101 Matrix Scientific

1H,2H,3H,4H,5H,7H-Furo[3,4-d]pyrimidine-2,4,7-trione

  • MDL number MFCD00702859
  • Cas Number : 4156-75-6
  • Molecular Weight : 168.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/118760.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/118760.html
Availability details for
Quantity
  • Availability
  • Add to Cart