MAT432343673 Matrix Scientific

2-(4-Trifluoromethyl phenyl)-ethenesulfonyl chloride

  • MDL number MFCD04973461
  • Cas Number : 929256-65-5
  • Molecular Weight : 270.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/120093.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/120093.html
Availability details for
Quantity
  • Availability
  • Add to Cart