MAT432343676 Matrix Scientific

2-(3,4-Difluorophenyl)-ethenesulfonyl chloride

  • MDL number MFCD04973465
  • Cas Number : 1158108-22-5
  • Molecular Weight : 238.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/120096.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/120096.html
Availability details for
Quantity
  • Availability
  • Add to Cart