MAT443486348 Matrix Scientific

2-(Bromomethyl)-3-chloro-5-(trifluoromethyl)pyridine

  • MDL number MFCD16607521
  • Cas Number : 1227502-50-2
  • Molecular Weight : 274.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Product Link https://www.matrixscientific.com/120290.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart