MAT443738081 Matrix Scientific

7-(2-Methylphenyl)thieno[3,2-d]pyrimidin-4(3H)-one

  • MDL number MFCD16639688
  • Cas Number : 1030120-83-2
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin US
Product Link https://www.matrixscientific.com/120445.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart