MAT443738398 Matrix Scientific

2-Fluoroacetyl chloride

  • MDL number MFCD00015738
  • Cas Number : 359-06-8
  • Molecular Weight : 96.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerate/argon
Country of Origin USA
Boiling point 70-71
Product Link http://www.matrixscientific.com/120763.html

Safety Information

hcodes Toxic/Corrosive/Stored under Argon/Moisture Sensitive/Refrigerate/Lachrymator
MSDS http://www.matrixscientific.com/120763.html
Availability details for
Quantity
  • Availability
  • Add to Cart