MAT447398437 Matrix Scientific

Olivetolic acid

  • MDL number MFCD28383644
  • Cas Number : 491-72-5
  • Molecular Weight : 224.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/121441.html

Safety Information

hcodes Irritant/Refrigerate
Availability details for
Quantity
  • Availability
  • Add to Cart