MAT448688780 Matrix Scientific

(2S,3S)-2,3-Dihydroxy-3-(phenylcarbamoyl)-propanoic acid

  • MDL number MFCD04038132
  • Cas Number : 206761-64-0
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Product Link http://www.matrixscientific.com/121963.html

Safety Information

MSDS http://www.matrixscientific.com/121963.html
Availability details for
Quantity
  • Availability
  • Add to Cart