MAT448689179 Matrix Scientific

Dapagliflozin

  • MDL number MFCD13182359
  • Cas Number : 461432-26-8
  • Molecular Weight : 408.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/122366.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/122366.html
Availability details for
Quantity
  • Availability
  • Add to Cart