MAT448902300 Matrix Scientific

(S)-Methyl 3-amino-6-chloro-5-(3-ethylpiperazin-1-yl)pyrazine-2-carboxylate

  • MDL number MFCD28359237
  • Cas Number : 906810-37-5
  • Molecular Weight : 299.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/125505.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/125505.html
Availability details for
Quantity
  • Availability
  • Add to Cart