MAT448902307 Matrix Scientific

3-Ethyl-1,2,4-oxadiazole-5-carboxylic acid, potassium salt

  • MDL number MFCD22548409
  • Cas Number : 944906-35-8
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036347.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036347.html
Availability details for
Quantity
  • Availability
  • Add to Cart