MAT448902308 Matrix Scientific

1-tert-Butyl 4-ethyl 4-phenylpiperidine-1,4-dicarboxylate

  • MDL number MFCD12032167
  • Cas Number : 1189118-20-4
  • Molecular Weight : 333.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/125479.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/125479.html
Availability details for
Quantity
  • Availability
  • Add to Cart