MAT448902309 Matrix Scientific

3-(2,4-Difluorophenoxy)pyrrolidine hydrochloride

  • MDL number MFCD28126193
  • Cas Number : 946715-62-4
  • Molecular Weight : 235.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023429.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023429.html
Availability details for
Quantity
  • Availability
  • Add to Cart