MAT448902315 Matrix Scientific

6-((2,2-Dimethyl-1,3-dioxolan-4-yl)methoxy)pyrazin-2-amine

  • MDL number MFCD15147079
  • Cas Number : 1219130-25-2
  • Molecular Weight : 225.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/125486.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/125486.html
Availability details for
Quantity
  • Availability
  • Add to Cart