MAT448902318 Matrix Scientific

(3,4-Dihydro-2H-pyrano[3,2-c]pyridin-7-yl)methanol hydrochloride

  • MDL number MFCD18802836
  • Cas Number : 959617-58-4
  • Molecular Weight : 201.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/125489.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/125489.html
Availability details for
Quantity
  • Availability
  • Add to Cart