MAT448902352 Matrix Scientific

4,4,5,5-Tetramethyl-2-(4-(1,1,1-trifluoro-2-methyl-propan-2-yl)phenyl)-1,3,2-dioxaborolane

  • MDL number MFCD28134483
  • Cas Number : 1432571-98-6
  • Molecular Weight : 314.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/125524.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/125524.html
Availability details for
Quantity
  • Availability
  • Add to Cart