MAT448902357 Matrix Scientific

Diethyl ketomalonate hydrate

  • MDL number MFCD28101749
  • Cas Number : 133318-40-8
  • Molecular Weight : 192.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57
Country of Origin USA
Product Link http://www.matrixscientific.com/125537.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/125537.html
Availability details for
Quantity
  • Availability
  • Add to Cart