MAT456777958 Matrix Scientific

Methyl 3-fluoro-6-(trifluoromethyl)picolinate

  • MDL number MFCD28714473
  • Cas Number : 1446509-52-9
  • Molecular Weight : 223.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/125662.html

Safety Information

MSDS http://www.matrixscientific.com/125662.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart