MAT456778123 Matrix Scientific

2,2-Dimethoxybutane

  • MDL number MFCD13185945
  • Cas Number : 3453-99-4
  • Molecular Weight : 118.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/125785.html

Safety Information

MSDS http://www.matrixscientific.com/125785.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart