MAT456989450 Matrix Scientific

4-Methyl-2-(1-pyrrolidinyl)-6,7-dihydro-5H-cyclopenta[D]pyrimidine

  • MDL number MFCD04125811
  • Cas Number : 866049-24-3
  • Molecular Weight : 203.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin US
Product Link https://www.matrixscientific.com/127765.html
Availability details for
Quantity
  • Availability
  • Add to Cart