MAT456989455 Matrix Scientific

1-Methyl-3-[(phenylsulfanyl)methyl]-1H-pyrazol-5-ol

  • MDL number MFCD00794867
  • Cas Number : 318234-24-1
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin US
Product Link http://www.matrixscientific.com/127772.html

Safety Information

MSDS http://www.matrixscientific.com/127772.html
Availability details for
Quantity
  • Availability
  • Add to Cart