MAT456989457 Matrix Scientific

1-(Benzyloxy)-1H-imidazole

  • MDL number MFCD01814553
  • Cas Number : 121779-19-9
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://www.matrixscientific.com/127774.html

Safety Information

MSDS http://www.matrixscientific.com/127774.html
Availability details for
Quantity
  • Availability
  • Add to Cart