MAT458026428 Matrix Scientific

Trifluoroacetaldehyde monohydrate, 75% aqueous solution

  • MDL number MFCD00167359
  • Cas Number : 421-53-4
  • Molecular Weight : 98.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/081758.html

Safety Information

MSDS http://www.matrixscientific.com/081758.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart