MAT458026433 Matrix Scientific

3-[3-(Trifluoromethyl)phenoxy]pyrrolidine hydrochloride

  • MDL number MFCD10567432
  • Cas Number : 21767-16-8
  • Molecular Weight : 267.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023390.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023390.html
Availability details for
Quantity
  • Availability
  • Add to Cart