MAT458028112 Matrix Scientific

4-[(Methylamino)sulphonyl]benzene boronic acid

  • MDL number MFCD06659864
  • Cas Number : 226396-31-2
  • Molecular Weight : 215.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Product Link https://www.matrixscientific.com/127756.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart