MAT458446653 Matrix Scientific

1-Methylpiperidin-3-one hydrochloride

  • MDL number MFCD18432588
  • Cas Number : 41511-85-7
  • Molecular Weight : 149.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/073504.html

Safety Information

MSDS http://www.matrixscientific.com/073504.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart