MAT458446716 Matrix Scientific

1-Benzyl-5-bromo-1H-1,2,4-triazole

  • MDL number MFCD21099399
  • Cas Number : 1352925-80-4
  • Molecular Weight : 238.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/128305.html

Safety Information

MSDS http://www.matrixscientific.com/128305.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart