MAT458446727 Matrix Scientific

1-Iodo-3-fluoropropane

  • MDL number MFCD07784197
  • Cas Number : 462-40-8
  • Molecular Weight : 187.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 34
Country of Origin US
Boiling point 68-69�mm
Product Link https://www.matrixscientific.com/128316.html

Safety Information

hcodes Flammable/Light Sensitive/Air Sensitive/Refrigerate
Availability details for
Quantity
  • Availability
  • Add to Cart