MAT458449063 Matrix Scientific

3-Methoxypropanoyl chloride

  • MDL number MFCD03424697
  • Cas Number : 4244-59-1
  • Molecular Weight : 122.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient/argon
Country of Origin USA
Boiling point 137-140°
Product Link http://www.matrixscientific.com/130620.html

Safety Information

MSDS http://www.matrixscientific.com/130620.html
pictograms Corrosive/Stored under Argon
Availability details for
Quantity
  • Availability
  • Add to Cart