MAT465533875 Matrix Scientific

Boc-alpha-methyl-L-phenylalanine

  • MDL number MFCD02682284
  • Cas Number : 111771-58-5
  • Molecular Weight : 279.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/041899.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041899.html
Availability details for
Quantity
  • Availability
  • Add to Cart