MAT465533885 Matrix Scientific

Boc-alpha-methyl-L-phenylalanine

  • MDL number MFCD02682673
  • Cas Number : 53940-88-8
  • Molecular Weight : 279.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/041898.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041898.html
Availability details for
Quantity
  • Availability
  • Add to Cart