MAT490082535 Matrix Scientific

1H-1,3-Benzodiazole hydrochloride

  • MDL number MFCD10676817
  • Cas Number : 18227-18-4
  • Molecular Weight : 154.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/133719.html

Safety Information

hcodes Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart