MAT490089240 Matrix Scientific

Pyrazolo[1,5-a]pyrimidin-6-amine

  • MDL number MFCD09701813
  • Cas Number : 1018125-39-7
  • Molecular Weight : 134.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/140230.html

Safety Information

MSDS http://www.matrixscientific.com/140230.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart