MAT490091739 Matrix Scientific

Methyl 2-bromo-3,4-difluorobenzoate

  • MDL number MFCD22490610
  • Cas Number : 1805523-36-7
  • Molecular Weight : 251.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/140950.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/140950.html
Availability details for
Quantity
  • Availability
  • Add to Cart