MAT491649945 Matrix Scientific

Dimethyl-(6-pyrrolidin-2-yl-pyridin-2-yl)-amine hydrochloride

  • MDL number MFCD29477423
  • Cas Number : 1909309-54-1
  • Molecular Weight : 227.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/068154.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/068154.html
Availability details for
Quantity
  • Availability
  • Add to Cart