MAT491649949 Matrix Scientific

(S)-(R)-1-((4-((5-Fluoro-2-methyl-1H-indol-6-yl)oxy)-5-methylpyrrolo-[2,1-f][1,2,4]triazin-6-yl)oxy)propan-2-yl 2-aminopropanoate

  • MDL number MFCD28142640
  • Cas Number : 649735-63-7
  • Molecular Weight : 441.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/143737.html

Safety Information

MSDS http://www.matrixscientific.com/143737.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart