MAT491651670 Matrix Scientific

4-Bromo-1-methyl-1H-imidazole-2-carbaldehyde

  • MDL number MFCD22420225
  • Cas Number : 79326-91-3
  • Molecular Weight : 189.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient/argon
Country of Origin USA
Product Link http://www.matrixscientific.com/145134.html

Safety Information

MSDS http://www.matrixscientific.com/145134.html
pictograms Irritant/Stored under Argon
Availability details for
Quantity
  • Availability
  • Add to Cart